Cyclo(7-aminoheptanoyl-Phe-D-Trp-Lys-Thr[Bzl])
Synonym(s):
Cyclosomatostatin
Empirical Formula (Hill Notation):
C44H57N7O6
CAS Number:
Molecular Weight:
779.97
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing