Skip to Content
Merck
Copper(I) cyanide 99%

Copper(I) cyanide

Copper(I) cyanide 99%
Synonym(s):
Cuprous cyanide, Copper(I) cyanide
Linear Formula:
CuCN
CAS Number:
Molecular Weight:
89.56

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing