
CL316243 Hydrate - CAS 138908-40-4 - Calbiochem

Synonym(s):
CL 316,243 hydrate, Disodium 5-[(2R)-2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]-1,3-benzodioxole-2,2-dicarboxylate hydrate
Empirical Formula (Hill Notation):
C20H18ClNNa2O7 · xH2O
CAS Number:
Molecular Weight:
465.79 (anhydrous basis)
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing