Skip to Content
Merck
Cholecalciferol analytical standard

Cholecalciferol

Cholecalciferol analytical standard
Synonym(s):
(+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique