Skip to Content
Merck
Chlorocyclohexane 99%

Chlorocyclohexane

Chlorocyclohexane 99%
Synonym(s):
Cyclohexyl chloride, Chlorocyclohexane
Empirical Formula (Hill Notation):
C6H11Cl
CAS Number:
Molecular Weight:
118.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing