Chlordiazepoxide
Synonym(s):
7-Chloro-N-methyl-5-phenyl-3H-1,4-benzodiazepin-2-amine 4-oxide
Empirical Formula (Hill Notation):
C16H14ClN3O
CAS Number:
Molecular Weight:
299.75
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing