Skip to Content
Merck
vibrant-m

Cephalexin Related Compound D

vibrant-m
Synonym(s):
3-Hydroxy-4-methylthiophen-2(5H)-one
Empirical Formula (Hill Notation):
C5H6O2S
CAS Number:
Molecular Weight:
130.16

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1