Bis(2-ethylhexyl)phthalate-3,4,5,6-d4
Synonym(s):
DEHP-3,4,5,6-d4, Deuterated DEHP, Deuterated bis(2-ethylhexyl)phthalate, Dioctyl phthalate-3,4,5,6-d4, Phthalic acid-3,4,5,6-d4 bis(2-ethylhexyl ester)
Empirical Formula (Hill Notation):
C24D4H34O4
CAS Number:
Molecular Weight:
394.58
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing