Skip to Content
Merck
Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt ≥90% (TLC), powder

Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt

Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt ≥90% (TLC), powder
Synonym(s):
Sulfosuccinimidyl 6-(biotinamido)hexanoate
Empirical Formula (Hill Notation):
C20H29N4NaO9S2
CAS Number:
Molecular Weight:
556.59

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1