Skip to Content
Merck
Bifenthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Bifenthrin

Bifenthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester
Empirical Formula (Hill Notation):
C23H22ClF3O2
CAS Number:
Molecular Weight:
422.87

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade