![Benzyltrimethylammonium chloride 97%](/deepweb/assets/sigmaaldrich/product/structures/383/208/a537bed1-d7a7-43db-b59a-d16ef7488827/640/a537bed1-d7a7-43db-b59a-d16ef7488827.png)
Benzyltrimethylammonium chloride
![Benzyltrimethylammonium chloride 97%](/deepweb/assets/sigmaaldrich/product/structures/383/208/a537bed1-d7a7-43db-b59a-d16ef7488827/640/a537bed1-d7a7-43db-b59a-d16ef7488827.png)
Synonym(s):
Benzyltrimethylammonium chloride
Linear Formula:
C6H5CH2(CH3)3NCl
CAS Number:
Molecular Weight:
185.69
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing