Benazepril Related Compound B
Synonym(s):
(3S)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride
Empirical Formula (Hill Notation):
C24H28N2O5 · HCl
CAS Number:
Molecular Weight:
460.95
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2