Skip to Content
Merck
Beauvericin ≥97% (HPLC)

Beauvericin

Beauvericin ≥97% (HPLC)
Synonym(s):
cyclo(D-α-Hydroxyisovaleryl-L-N-methyl-Phe)3
Empirical Formula (Hill Notation):
C45H57N3O9
CAS Number:
Molecular Weight:
783.95

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing