Skip to Content
Merck
Auraptene ≥98% (HPLC)

Auraptene

Auraptene ≥98% (HPLC)
Synonym(s):
7-Geranyloxycoumarin, 7-[[(2E)-3,7-dimethyl-2,6-octadien-1-yl]oxy]-2H-1-benzopyran-2-one, Aurapten
Empirical Formula (Hill Notation):
C19H22O3
CAS Number:
Molecular Weight:
298.38

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing