Skip to Content
Merck
vibrant-m

Apramycin

vibrant-m
Synonym(s):
O-4-Amino-4-deoxy-α-D-glucopyranosyl-(1→8)-O-(8R)-2-amino-2,3,7-trideoxy-7-(methylamino)-D-glycero-α-D-allo-octodialdo-1,5:8,4-dipyranosyl-(1→4)-2-deoxy-D-streptamine
Empirical Formula (Hill Notation):
C21H41N5O11
CAS Number:
Molecular Weight:
539.58
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing