Anti-ZC3H12A antibody produced in rabbit
Synonym(s):
Anti-FLJ23231, Anti-MCPIP1, Anti-zinc finger CCCH-type containing 12A, FLJ23231, MCPIP, MCPIP1, ZC3H12A, Anti-MCP-1-induced protein, Anti-MCPIP, Anti-Zinc finger CCCH-type containing 12A
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing