
Anti-Swine H1N1 Hemagglutinin antibody produced in rabbit

Synonym(s):
Anti-HA, Anti-S-OIV A Hemagglutinin, Anti-Swine flu H1, Anti-Swine-Origin Influenza A (H1N1) Hemagglutinin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing