![Ammonium monovanadate GR for analysis Reag. Ph Eur](/deepweb/assets/sigmaaldrich/product/images/350/751/c4f466f3-0750-4cb4-af8f-5942eba5d12f/640/c4f466f3-0750-4cb4-af8f-5942eba5d12f.jpg)
Ammonium monovanadate
![Ammonium monovanadate GR for analysis Reag. Ph Eur](/deepweb/assets/sigmaaldrich/product/images/350/751/c4f466f3-0750-4cb4-af8f-5942eba5d12f/640/c4f466f3-0750-4cb4-af8f-5942eba5d12f.jpg)
Synonym(s):
Ammonium monovanadate
Empirical Formula (Hill Notation):
H4NO3V
CAS Number:
Molecular Weight:
116.98
Products
Formula Weight
Physical Form
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing