Skip to Content
Merck
Amitriptyline-d3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Amitriptyline-d3 hydrochloride solution

Amitriptyline-d3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Amitriptyline-d3 hydrochloride solution
Empirical Formula (Hill Notation):
C20D3H21ClN
CAS Number:
Molecular Weight:
316.88
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique