![Amitraz Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/265/042/ae022b5e-d2dc-4d77-b395-b045219a0e59/640/ae022b5e-d2dc-4d77-b395-b045219a0e59.png)
AMITRAZ RELATED COMPOUND A
![Amitraz Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/265/042/ae022b5e-d2dc-4d77-b395-b045219a0e59/640/ae022b5e-d2dc-4d77-b395-b045219a0e59.png)
Synonym(s):
N-(2,4-Dimethylphenyl)formamide
Linear Formula:
(CH3)2C6H3NHCHO
CAS Number:
Molecular Weight:
149.19
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2