Amifostine thiol
Synonym(s):
WR-1065, 2-[(3-Aminopropyl)amino]ethanethiol dihydrochloride, WR-1065 dihydrochloride
Empirical Formula (Hill Notation):
C5H14N2S · 2HCl
CAS Number:
Molecular Weight:
207.16
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2