
Aluminium chloride

Synonym(s):
Aluminium chloride
Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
Products
Brand
Formula Weight
Physical Form
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing