Alprenolol hydrochloride
Synonym(s):
1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride
Empirical Formula (Hill Notation):
C15H23NO2 · HCl
CAS Number:
Molecular Weight:
285.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing