Acid Fuchsin calcium salt
Synonym(s):
Acid Magenta, Acid Rubin, Fuchsin S, SS, SS, ST, or SIII, Rubin S
Linear Formula:
C20H17N3O9S3Ca
CAS Number:
Molecular Weight:
579.64
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing