6-Hydroxymelatonin
Synonym(s):
3-(N-Acetylaminoethyl)-6-hydroxy-5-methoxyindole
Empirical Formula (Hill Notation):
C13H16N2O3
CAS Number:
Molecular Weight:
248.28
Products
Product Category
Brand
Formula Weight
Gene Alias
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing