Skip to Content
Merck
5-Nitro-2-(3-phenylpropylamino)benzoic acid ≥98%

5-Nitro-2-(3-phenylpropylamino)benzoic acid

5-Nitro-2-(3-phenylpropylamino)benzoic acid ≥98%
Synonym(s):
NPPB, 5-Nitro-2-(3-phenylpropylamino)benzoic Acid
Empirical Formula (Hill Notation):
C16H16N2O4
CAS Number:
Molecular Weight:
300.31

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment