![5-Bromo-2′-deoxyuridine >97%, crystalline solid, pkg of 1 g](/deepweb/assets/sigmaaldrich/product/structures/113/596/8ee7ed11-aa72-4938-94da-a66a4ff4f260/640/8ee7ed11-aa72-4938-94da-a66a4ff4f260.png)
5-Bromo-2′-deoxyuridine
![5-Bromo-2′-deoxyuridine >97%, crystalline solid, pkg of 1 g](/deepweb/assets/sigmaaldrich/product/structures/113/596/8ee7ed11-aa72-4938-94da-a66a4ff4f260/640/8ee7ed11-aa72-4938-94da-a66a4ff4f260.png)
Synonym(s):
5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR, 5-Bromo-2′-deoxyuridine
Empirical Formula (Hill Notation):
C9H11BrN2O5
CAS Number:
Molecular Weight:
307.10
Products
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing