4-Nitrophenyl acetate
Synonym(s):
Acetic acid 4-nitrophenyl ester, 4-Nitrophenyl acetate
Linear Formula:
CH3CO2C6H4NO2
CAS Number:
Molecular Weight:
181.15
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing