
4-Methoxy-2(5H)-furanone

Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing