Skip to Content
Merck
4′-Hydroxydiclofenac VETRANAL®, analytical standard

4′-Hydroxydiclofenac

4′-Hydroxydiclofenac VETRANAL®, analytical standard
Synonym(s):
4′-Hydroxy Diclofenac, [2-(2,6-Dichloro-4-hydroxy-phenylamino)phenyl]acetic acid
Empirical Formula (Hill Notation):
C14H11Cl2NO3
CAS Number:
Molecular Weight:
312.15

Products

Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing