Skip to Content
Merck
4-(Diethoxymethyl)benzaldehyde 97%

4-(Diethoxymethyl)benzaldehyde

4-(Diethoxymethyl)benzaldehyde 97%
Synonym(s):
Terephthalaldehyde diethyl acetal
Linear Formula:
(C2H5O)2CHC6H4CHO
CAS Number:
Molecular Weight:
208.25

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing