![4-Bromobenzylsulfonyl chloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/127/457/956d1371-93d5-4a2b-8121-fd89c72ee361/640/956d1371-93d5-4a2b-8121-fd89c72ee361.png)
4-Bromobenzylsulfonyl chloride
![4-Bromobenzylsulfonyl chloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/127/457/956d1371-93d5-4a2b-8121-fd89c72ee361/640/956d1371-93d5-4a2b-8121-fd89c72ee361.png)
Empirical Formula (Hill Notation):
C7H6BrClO2S
Molecular Weight:
269.54
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing