Skip to Content
Merck
4-Aminobiphenyl analytical standard

4-Aminobiphenyl

4-Aminobiphenyl analytical standard
Synonym(s):
4-Aminodiphenyl, 4-Biphenylamine, 4-Biphenylylamine, 4-Phenylaniline, Biphenyl-4-ylamine, NSC 7660, Xenylamine
Linear Formula:
C6H5C6H4NH2
CAS Number:
Molecular Weight:
169.22
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing