Skip to Content
Merck
3-sn-Phosphatidic acid sodium salt from egg yolk lecithin ≥98%

3-sn-Phosphatidic acid sodium salt from egg yolk lecithin

3-sn-Phosphatidic acid sodium salt from egg yolk lecithin ≥98%
Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphate sodium salt, L-Ī±-Phosphatidic acid sodium salt, PA

Products

Product Category
Brand
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing