Skip to Content
Merck
3-Octanol analytical standard

3-Octanol

3-Octanol analytical standard
Synonym(s):
Ethyl pentyl carbinol
Linear Formula:
CH3(CH2)4CH(OH)CH2CH3
CAS Number:
Molecular Weight:
130.23
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing