![3-Chloro-1H-pyrazolo[3,4-b]pyridine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/381/424/670964a4-3d9a-4dc2-966b-ede1f945daf0/640/670964a4-3d9a-4dc2-966b-ede1f945daf0.png)
3-Chloro-1H-pyrazolo[3,4-b]pyridine
![3-Chloro-1H-pyrazolo[3,4-b]pyridine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/381/424/670964a4-3d9a-4dc2-966b-ede1f945daf0/640/670964a4-3d9a-4dc2-966b-ede1f945daf0.png)
Empirical Formula (Hill Notation):
C6H4ClN3
CAS Number:
Molecular Weight:
153.57
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing