Skip to Content
Merck
3-Buten-2-one contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%

3-Buten-2-one

3-Buten-2-one contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%
Synonym(s):
Methyl vinyl ketone, Vinyl methyl ketone
Linear Formula:
CH2=CHCOCH3
CAS Number:
Molecular Weight:
70.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1