2-Methylglutaric acid
Linear Formula:
HOOCCH2CH2CH(CH3)COOH
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing