2-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine
Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing