
2-Methoxyethoxymethyl chloride

Synonym(s):
β-Methoxyethoxymethyl chloride, MEM-chloride
Linear Formula:
CH3OCH2CH2OCH2Cl
CAS Number:
Molecular Weight:
124.57
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1