2-Hydroxybenzyl alcohol
Synonym(s):
Salicyl alcohol, Saligenin, 2-Hydroxybenzyl alcohol
Linear Formula:
2-(OH)C6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing