![2-furaldehyde certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/363/175/728405ea-3163-45a9-9031-23c5c0fb8289/640/728405ea-3163-45a9-9031-23c5c0fb8289.png)
2-furaldehyde
![2-furaldehyde certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/363/175/728405ea-3163-45a9-9031-23c5c0fb8289/640/728405ea-3163-45a9-9031-23c5c0fb8289.png)
Synonym(s):
Furfural, 2-Furaldehyde, Furan-2-carboxaldehyde
Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing