2-(Ethylamino)ethanol
Synonym(s):
N-Ethylethanolamine, 2-(Ethylamino)ethanol
Linear Formula:
C2H5NHCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing