![2′-Deoxyguanosine monohydrate 99-100%](/deepweb/assets/sigmaaldrich/product/structures/144/945/68a140c1-dbf8-4faa-b1c5-ed6344299e7c/640/68a140c1-dbf8-4faa-b1c5-ed6344299e7c.png)
2′-Deoxyguanosine monohydrate
![2′-Deoxyguanosine monohydrate 99-100%](/deepweb/assets/sigmaaldrich/product/structures/144/945/68a140c1-dbf8-4faa-b1c5-ed6344299e7c/640/68a140c1-dbf8-4faa-b1c5-ed6344299e7c.png)
Synonym(s):
9-(2-Deoxy-β-D-ribofuranosyl)guanine, Guanine-2′-deoxyriboside
Empirical Formula (Hill Notation):
C10H13N5O4 · H2O
CAS Number:
Molecular Weight:
285.26
Products
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing