Skip to Content
Merck
25-Hydroxyvitamin D2 solution 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

25-Hydroxyvitamin D2 solution

25-Hydroxyvitamin D2 solution 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C28H44O2
CAS Number:
Molecular Weight:
412.65
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade