![2-[4-(Butylamino)benzoyl]oxy-N,N-dimethylethanamine-N-oxide hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/239/775/7db11b19-cc5b-4461-855d-187aa5a2f421/640/7db11b19-cc5b-4461-855d-187aa5a2f421.png)
2-[4-(Butylamino)benzoyl]oxy-N,N-dimethylethanamine-N-oxide hydrochloride
![2-[4-(Butylamino)benzoyl]oxy-N,N-dimethylethanamine-N-oxide hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/239/775/7db11b19-cc5b-4461-855d-187aa5a2f421/640/7db11b19-cc5b-4461-855d-187aa5a2f421.png)
Synonym(s):
Tetracaine N-oxide hydrochloride
Empirical Formula (Hill Notation):
C15H24N2O3 · HCl
Molecular Weight:
316.82
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1