![(−)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane 98%](/deepweb/assets/sigmaaldrich/product/structures/270/536/5a7f3259-4155-4821-b0a1-4b02883fe433/640/5a7f3259-4155-4821-b0a1-4b02883fe433.png)
(−)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
![(−)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane 98%](/deepweb/assets/sigmaaldrich/product/structures/270/536/5a7f3259-4155-4821-b0a1-4b02883fe433/640/5a7f3259-4155-4821-b0a1-4b02883fe433.png)
Synonym(s):
(−)-1,4-Bis(diphenylphosphino)-1,4-dideoxy-2,3-O-isopropylidene-L-threitol, (4R,5R)-4,5-Bis(diphenylphosphino-methyl)-2,2-dimethyl-1,3-dioxolane, (4R-trans)-[(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene)]bis(diphenylphosphine), (R,R)-DIOP
Empirical Formula (Hill Notation):
C31H32O2P2
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing