Skip to Content
Merck
20-Hydroxyecdysone ≥93% (HPLC), powder

20-Hydroxyecdysone

20-Hydroxyecdysone ≥93% (HPLC), powder
Synonym(s):
β-Ecdysone, 2β,3β,14α,20β,22,25-Hexahydroxy-7-cholesten-6-one, Ecdysterone, Insect moulting hormone, Polypodine A
Empirical Formula (Hill Notation):
C27H44O7
CAS Number:
Molecular Weight:
480.63

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing