Skip to Content
Merck
1(S),9(R)-(−)-Bicuculline methiodide ≥95.0% (HPCE)

1(S),9(R)-(−)-Bicuculline methiodide

1(S),9(R)-(−)-Bicuculline methiodide ≥95.0% (HPCE)
Synonym(s):
(5S)-5-[(6R)-6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl]-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium iodide
Empirical Formula (Hill Notation):
C21H20INO6
CAS Number:
Molecular Weight:
509.29

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing