![1-Methylhistamine dihydrochloride ≥98% (TLC), powder](/deepweb/assets/sigmaaldrich/product/structures/237/063/13514ea1-c733-47db-a60e-7acd05526024/640/13514ea1-c733-47db-a60e-7acd05526024.png)
1-Methylhistamine dihydrochloride
![1-Methylhistamine dihydrochloride ≥98% (TLC), powder](/deepweb/assets/sigmaaldrich/product/structures/237/063/13514ea1-c733-47db-a60e-7acd05526024/640/13514ea1-c733-47db-a60e-7acd05526024.png)
Synonym(s):
1-Methyl-4-(β-aminoethyl)imidazole dihydrochloride
Empirical Formula (Hill Notation):
C6H11N3 · 2HCl
CAS Number:
Molecular Weight:
198.09
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing